Gene Transcripts

Gene Symbol
TSPAN5
Entrez Gene ID
10098
Gene Name
tetraspanin 5
Gene Aliases
NET-4, NET4, TM4SF9, TSPAN-5
Chromosome Location
Chr. 4: 98470367 - 98658661 on Build GRCh38
UniGene ID
Hs.118118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005723.3 NP_005714.2 5 917
XM_005262680.1 XP_005262737.1 4 770
GenBank mRNA AF065389.1 - 5 829
AK126702.1 - 2 3556
AK295385.1 - 5 511
AK315723.1 - 5 884
BC009704.1 - 5 815 3895933
CR541809.1 - 5 482
CR541829.1 - 5 482

View Full Product Details