Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001023567.4 | NP_001018861.3 | 16 | 2738 | |
NR_027410.1 | 24 | 4986 | |||
XM_011521596.1 | XP_011519898.1 | 23 | 3583 | ||
XM_011521597.1 | XP_011519899.1 | 22 | 3534 | ||
XM_017022210.1 | XP_016877699.1 | 24 | 3887 | ||
XM_017022211.1 | XP_016877700.1 | 24 | 3656 | ||
XM_017022212.1 | XP_016877701.1 | 23 | 4210 | ||
XM_017022213.1 | XP_016877702.1 | 23 | 3612 | ||
XM_017022214.1 | XP_016877703.1 | 24 | 3884 | ||
XM_017022215.1 | XP_016877704.1 | 24 | 4409 | ||
XM_017022216.1 | XP_016877705.1 | 23 | 5007 | ||
XM_017022217.1 | XP_016877706.1 | 22 | 4929 | ||
XM_017022218.1 | XP_016877707.1 | 24 | 5036 | ||
GenBank mRNA | BX647673.1 | - | 23 | 4954 |