Gene Transcripts

Gene Symbol
RPS14P3
Entrez Gene ID
644068
Gene Name
ribosomal protein S14 pseudogene 3
Gene Aliases
RPS14_1_16
Chromosome Location
Chr. 1: 19607807 - 19608644 on Build GRCh38
UniGene ID
Hs.660760
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_077246.1 1 441
GenBank mRNA BC072682.1 - 1 441
BQ930453.1 - 1 443
BU176905.1 - 1 409

View Full Product Details