Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000842.4 | NP_000833.1 | 2 | 225 | |
NM_001143831.2 | NP_001137303.1 | 1 | 27 | ||
XM_006718828.3 | XP_006718891.1 | 2 | 223 | ||
XM_011542792.1 | XP_011541094.1 | 2 | 225 | ||
XM_017017627.1 | XP_016873116.1 | 2 | 223 | ||
GenBank mRNA | AK291362.1 | - | 2 | 195 | |
BX282658.1 | - | 2 | 197 | ||
DA316590.1 | - | 2 | 226 | ||
DR002861.1 | - | 2 | 225 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_027044.1 | 2 | 215 | ||
GenBank mRNA | AK294678.1 | - | 2 | 215 | |
BC141819.1 | - | 2 | 123 | ||
BC142657.1 | - | 2 | 123 |