Gene Transcripts
- Gene Symbol
- SQLE
- Entrez Gene ID
- 6713
- Gene Name
- squalene epoxidase
- Gene Aliases
- Chromosome Location
- Chr. 8: 124998478 - 125022283 on Build GRCh38
- UniGene ID
- Hs.71465
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003129.3 |
NP_003120.2
|
9, 10 |
2378 |
|
XM_011517246.2 |
XP_011515548.1
|
9, 10 |
1476 |
|
GenBank mRNA |
AF098865.1 |
- |
9, 10 |
1607 |
|
AK225085.1 |
- |
9, 10 |
1668 |
|
AK295302.1 |
- |
9, 10 |
1278 |
|
AK301792.1 |
- |
9, 10 |
1270 |
|
AK313384.1 |
- |
9, 10 |
1496 |
|
BC017033.1 |
- |
9, 10 |
1610 |
3846346 |
BX647400.1 |
- |
9, 10 |
2312 |
|
D78129.1 |
- |
7, 8 |
896 |
|
D78130.1 |
- |
9, 10 |
1715 |
|
View Full Product Details