Gene Transcripts
- Gene Symbol
- FAM25C
- Entrez Gene ID
- 644054
- Gene Name
- family with sequence similarity 25 member C
- Gene Aliases
- bA164N7.4
- Chromosome Location
- Chr. 10: 47995322 - 47999791 on Build GRCh38
- UniGene ID
- Hs.645212
- Species
- Homo sapiens
- Gene Symbol
- FAM25BP
- Entrez Gene ID
- 100132929
- Gene Name
- protein FAM25
- Gene Aliases
- FAM25A, FAM25B, FAM25C, FAM25G
- Chromosome Location
- Chr. 10: 46369087 - 46373563 on Build GRCh38
- UniGene ID
- Hs.645212
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104039.1 |
|
3 |
223 |
|
GenBank mRNA |
BC146968.1 |
- |
3 |
220 |
|
- Gene Symbol
- FAM25G
- Entrez Gene ID
- 100133093
- Gene Name
- family with sequence similarity 25 member G
- Gene Aliases
- FAM25A, FAM25B, FAM25C, bA301J7.4
- Chromosome Location
- Chr. 10: 47487236 - 47491700 on Build GRCh38
- UniGene ID
- Hs.723078
- Species
- Homo sapiens
View Full Product Details