Gene Transcripts

Gene Symbol
RABEPK
Entrez Gene ID
10244
Gene Name
Rab9 effector protein with kelch motifs
Gene Aliases
RAB9P40, bA65N13.1, p40
Chromosome Location
Chr. 9: 125200493 - 125234159 on Build GRCh38
UniGene ID
Hs.19012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174152.1 NP_001167623.1 9 1016
NM_001174153.1 NP_001167624.1 8 863
NM_005833.3 NP_005824.2 8 1177
XM_005251640.4 XP_005251697.1 8 1089
XM_005251641.4 XP_005251698.1 7 944
XM_005251642.4 XP_005251699.1 7 935
XM_005251644.4 XP_005251701.1 6 785
XM_011518120.2 XP_011516422.1 8 1251
XM_017014177.1 XP_016869666.1 9 1001
XM_017014178.1 XP_016869667.1 8 903
XM_017014179.1 XP_016869668.1 7 976
XM_017014180.1 XP_016869669.1 7 750
XM_017014181.1 XP_016869670.1 6 861
XM_017014182.1 XP_016869671.1 7 686
XM_017014183.1 XP_016869672.1 7 1182
GenBank mRNA AK290768.1 - 8 1021
BC000503.2 - 8 1154 2821471
BC047023.1 - 8 843 5396980
BC053541.1 - 8 986 5724458
BC065725.1 - 9 1019 4102355
CR456837.1 - 7 847
CR541851.1 - 7 847
KU178507.1 - 7 847
KU178508.1 - 7 760
KU178509.1 - 6 698
KU178510.1 - 6 694
KU178511.1 - 5 532
Z97074.1 - 9 996

View Full Product Details