Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001023567.4 | NP_001018861.3 | 2 | 224 | |
NR_027410.1 | 11 | 1381 | |||
XM_011521596.1 | XP_011519898.1 | 9 | 1069 | ||
XM_011521597.1 | XP_011519899.1 | 8 | 1020 | ||
XM_017022210.1 | XP_016877699.1 | 10 | 1373 | ||
XM_017022211.1 | XP_016877700.1 | 10 | 1142 | ||
XM_017022212.1 | XP_016877701.1 | 9 | 1696 | ||
XM_017022213.1 | XP_016877702.1 | 9 | 1098 | ||
XM_017022214.1 | XP_016877703.1 | 10 | 1373 | ||
XM_017022215.1 | XP_016877704.1 | 10 | 1373 | ||
XM_017022216.1 | XP_016877705.1 | 10 | 1402 | ||
XM_017022217.1 | XP_016877706.1 | 9 | 1324 | ||
XM_017022218.1 | XP_016877707.1 | 10 | 2000 | ||
GenBank mRNA | AK097551.1 | - | 2 | 213 | |
BC143254.1 | - | 2 | 170 | 9051756 | |
BX647673.1 | - | 10 | 1350 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181077.3 | NP_851422.1 | 2 | 224 | |
NR_027409.1 | 11 | 1395 | |||
XM_006720440.2 | XP_006720503.1 | 2 | 192 | ||
XM_017022018.1 | XP_016877507.1 | 9 | 1176 | ||
XM_017022019.1 | XP_016877508.1 | 8 | 872 | ||
GenBank mRNA | AK123221.1 | - | 7 | 740 | |
BC032774.1 | - | 2 | 126 | ||
BC093673.1 | - | 2 | 170 | 7939518 | |
BC104800.1 | - | 2 | 170 | 8143803 | |
BX648160.1 | - | 10 | 1350 | ||
BX648575.1 | - | 10 | 1102 | ||
CR749361.1 | - | 3 | 286 |