Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001047160.2 | NP_001040625.1 | 9 | 1085 | |
NM_005863.4 | NP_005854.2 | 7 | 996 | ||
NR_073040.1 | 7 | 1029 | |||
GenBank mRNA | AB209852.1 | - | 9 | 1031 | |
AJ010046.1 | - | 7 | 929 | ||
AK024919.1 | - | 7 | 936 | ||
AK294244.1 | - | 3 | 462 | ||
AK299335.1 | - | 9 | 1041 | ||
AK302687.1 | - | 9 | 867 | ||
AK304028.1 | - | 7 | 971 | ||
AK315566.1 | - | 7 | 938 | ||
BC010285.1 | - | 7 | 927 | 3048980 | |
BC053553.1 | - | 9 | 1037 | 6137683 | |
BX537509.1 | - | 4 | 3209 | ||
U02081.1 | - | 5 | 517 |