Gene Transcripts
- Gene Symbol
- Ehbp1l1
- Entrez Gene ID
- 114601
- Gene Name
- EH domain binding protein 1-like 1
- Gene Aliases
- G430002G23Rik
- Chromosome Location
- Chr. 19: 5707369 - 5726431 on Build GRCm38
- UniGene ID
- Mm.210447
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114595.1 |
NP_001108067.1
|
9, 10 |
1205 |
|
NM_001114596.1 |
NP_001108068.1
|
10, 11 |
1241 |
|
NM_001114597.1 |
NP_001108069.1
|
10, 11 |
4073 |
|
NM_053252.3 |
NP_444482.2
|
10, 11 |
1550 |
|
GenBank mRNA |
AF305087.1 |
- |
10, 11 |
4029 |
|
AF305088.1 |
- |
10, 11 |
1503 |
|
AF305089.1 |
- |
9, 10 |
1158 |
|
AF305090.1 |
- |
10, 11 |
1194 |
|
AK087817.1 |
- |
6, 7 |
792 |
|
AK089917.1 |
- |
9, 10 |
1166 |
|
AK169974.1 |
- |
10, 11 |
1753 |
|
AK194109.1 |
- |
4, 5 |
366 |
|
BC004660.1 |
- |
10, 11 |
1499 |
3497913 |
View Full Product Details