Gene Transcripts
- Gene Symbol
- Slc7a2
- Entrez Gene ID
- 11988
- Gene Name
- solute carrier family 7 (cationic amino acid transporter, y+ system), member 2
- Gene Aliases
- 20.5, AI158848, Atrc2, CAT-2, Cat2, Tea
- Chromosome Location
- Chr. 8: 40862367 - 40922070 on Build GRCm38
- UniGene ID
- Mm.4676
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001044740.2 |
NP_001038205.1
|
3, 4 |
743 |
|
NM_007514.3 |
NP_031540.2
|
3, 4 |
743 |
|
GenBank mRNA |
AK154621.1 |
- |
3, 4 |
717 |
|
AK155558.1 |
- |
3, 4 |
718 |
|
AK171369.1 |
- |
3, 4 |
715 |
|
BC021653.1 |
- |
3, 4 |
738 |
|
BC127082.1 |
- |
3, 4 |
659 |
40111372 |
CN710433.1 |
- |
2, 3 |
350 |
|
DQ086834.1 |
- |
3, 4 |
698 |
|
L03290.1 |
- |
2, 3 |
545 |
|
L11600.1 |
- |
2, 3 |
545 |
|
L29006.1 |
- |
2, 3 |
1068 |
|
M62838.1 |
- |
3, 4 |
656 |
|
View Full Product Details