Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009968.3 | NP_034098.1 | 3 | 792 | |
GenBank mRNA | AK098082.1 | - | 3 | 276 | |
AK133820.1 | - | 3 | 793 | ||
AK134281.1 | - | 3 | 277 | ||
BC043076.1 | - | 3 | 253 | 6404996 | |
S70056.1 | - | 3 | 304 |