Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005886.2 | NP_005877.2 | 7 | 878 | |
XM_005255772.4 | XP_005255829.1 | 7 | 908 | ||
XM_006721121.3 | XP_006721184.1 | 7 | 909 | ||
XM_006721122.2 | XP_006721185.1 | 7 | 1033 | ||
XM_006721123.3 | XP_006721186.1 | 7 | 908 | ||
XM_011522810.2 | XP_011521112.1 | 7 | 1547 | ||
XM_017022860.1 | XP_016878349.1 | 7 | 850 | ||
XM_017022861.1 | XP_016878350.1 | 7 | 1033 | ||
XM_017022862.1 | XP_016878351.1 | 7 | 1547 | ||
XM_017022863.1 | XP_016878352.1 | 7 | 1047 | ||
XM_017022864.1 | XP_016878353.1 | 7 | 1547 | ||
GenBank mRNA | AB209250.1 | - | 7 | 1017 | |
AF052432.1 | - | 6 | 544 | ||
BC001353.2 | - | 6 | 527 | 2964632 | |
BT007022.1 | - | 6 | 504 | ||
CR456762.1 | - | 6 | 504 |