Gene Transcripts

Gene Symbol
Gng8
Entrez Gene ID
14709
Gene Name
guanine nucleotide binding protein (G protein), gamma 8
Gene Aliases
G(y)8
Chromosome Location
Chr. 7: 16891737 - 16895451 on Build GRCm38
UniGene ID
Mm.103770
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010320.3 NP_034450.1 4 824
GenBank mRNA AK144609.1 - 4 825

View Full Product Details