Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006049.2 | NP_006040.1 | 1, 2 | 166 | |
GenBank mRNA | AF093593.1 | - | 1, 2 | 89 | |
AK024763.1 | - | 1, 2 | 138 | ||
AK312018.1 | - | 1, 2 | 138 | ||
CD685245.1 | - | 1, 2 | 203 | ||
CN347010.1 | - | 1, 2 | 88 | ||
DB037177.1 | - | 1, 2 | 98 |