Gene Transcripts

Gene Symbol
Il12a
Entrez Gene ID
16159
Gene Name
interleukin 12a
Gene Aliases
IL-12p35, Il-12a, Ll12a, p35
Chromosome Location
Chr. 3: 68690644 - 68698550 on Build GRCm38
UniGene ID
Mm.103783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF128210.1 - 2 67
AF128211.1 - 2 67
AF128212.1 - 2 67
AF128213.1 - 2 67
BC116851.1 - 2 71
BC116855.1 - 2 71
M86672.1 - 2 169

View Full Product Details