Gene Transcripts

Gene Symbol
Pld1
Entrez Gene ID
18805
Gene Name
phospholipase D1
Gene Aliases
AA536939, C85393, Pld1a, Pld1b
Chromosome Location
Chr. 3: 27938680 - 28133362 on Build GRCm38
UniGene ID
Mm.212039
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164056.1 NP_001157528.1 23, 24 2742
NM_008875.4 NP_032901.2 22, 23 2687
GenBank mRNA AK136111.1 - 17, 18 1925
BC026625.1 - 3, 4 202
BC068144.1 - 22, 23 2544 30532470
U87868.1 - 23, 24 2725

View Full Product Details