Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001036740.2 | NP_001031817.1 | 2 | 192 | |
NM_146184.4 | NP_666296.1 | 1 | 584 | ||
GenBank mRNA | AK042388.1 | - | 1 | 567 | |
AK154220.1 | - | 179 | |||
AK154901.1 | - | 2 | 148 | ||
BC025206.1 | - | 2 | 178 | 5037295 | |
BY320287.1 | - | 2 | 176 |