Gene Transcripts

Gene Symbol
Hist1h2aa
Entrez Gene ID
319163
Gene Name
histone cluster 1, H2aa
Gene Aliases
Th2a
Chromosome Location
Chr. 13: 23934462 - 23934913 on Build GRCm38
UniGene ID
Mm.377876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175658.2 NP_783589.1 1 68
GenBank mRNA BC107354.1 - 1 68
BC107355.1 - 1 68 40055860

View Full Product Details