Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181548.2 | NP_853526.1 | 2 | 622 | |
GenBank mRNA | AB093573.1 | - | 2 | 622 | |
AK141212.1 | - | 2 | 516 | ||
BC119277.1 | - | 2 | 538 | 8734026 | |
BC119281.1 | - | 2 | 538 | 8734030 |