Gene Transcripts
- Gene Symbol
- Igh-VJ558
- Entrez Gene ID
- 16061
- Gene Name
- immunoglobulin heavy chain (J558 family)
- Gene Aliases
- AI893585
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.246497
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF296427.1 |
- |
1 |
145 |
|
View Full Product Details