Gene Transcripts

Gene Symbol
Cramp1l
Entrez Gene ID
57354
Gene Name
Crm, cramped-like (Drosophila)
Gene Aliases
5830477H08Rik, AI256853, AI790734, Tce4, mKIAA1426
Chromosome Location
Chr. 17: 24961226 - 25015846 on Build GRCm38
UniGene ID
Mm.328854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020608.3 NP_065633.2 3 622
GenBank mRNA AK089393.1 - 3 872
AK147411.1 - 3 623
AK147449.1 - 3 623

View Full Product Details