Gene Transcripts

Gene Symbol
Atp13a2
Entrez Gene ID
74772
Gene Name
ATPase type 13A2
Gene Aliases
1110012E06Rik, AA589443
Chromosome Location
Chr. 4: 140986865 - 141007701 on Build GRCm38
UniGene ID
Mm.205625
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164366.1 NP_001157838.1 20 2312
NM_029097.2 NP_083373.2 20 2312
GenBank mRNA AK003623.1 - 4 377
AK171554.1 - 19 2567

View Full Product Details