Gene Transcripts

Gene Symbol
Rbfa
Entrez Gene ID
68731
Gene Name
ribosome binding factor A
Gene Aliases
1110032A13Rik, AI595940
Chromosome Location
Chr. 18: 80192264 - 80200619 on Build GRCm38
UniGene ID
Mm.186936
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199197.1 NP_954667.1 2, 3 461
GenBank mRNA AK004019.1 - 2, 3 480
AK145807.1 - 2, 3 500
BC064066.1 - 2, 3 461 935634

View Full Product Details