Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027768.3 | NP_082044.1 | 3 | 288 | |
GenBank mRNA | AK017258.1 | - | 3 | 289 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001085544.1 | NP_001079013.1 | 3 | 288 |