Gene Transcripts

Gene Symbol
SSSCA1
Entrez Gene ID
10534
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
Gene Aliases
p27
Chromosome Location
Chr. 11: 65570459 - 65571888 on Build GRCh38
UniGene ID
Hs.25723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303024.1 NP_001289953.1 2 322
NM_006396.2 NP_006387.1 3 229
GenBank mRNA AB001740.1 - 3 216
BC014791.2 - 3 215 3857068
BP351453.1 - 3 230
DR759640.1 - 2 289

View Full Product Details