Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014262.4 | NP_055077.2 | 7, 8 | 1335 | |
GenBank mRNA | AJ430349.1 | - | 8, 9 | 1564 | |
AK025976.1 | - | 7, 8 | 1302 | ||
BC017217.2 | - | 7, 8 | 876 | ||
BC067251.1 | - | 7, 8 | 1033 | ||
BC080630.1 | - | 7, 8 | 1108 | ||
U47926.1 | - | 6, 7 | 800 |