Gene Transcripts

Gene Symbol
Higd1b
Entrez Gene ID
75689
Gene Name
HIG1 domain family, member 1B
Gene Aliases
2310056K19Rik
Chromosome Location
Chr. 11: 102835756 - 102844133 on Build GRCm38
UniGene ID
Mm.44408
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080846.1 NP_543122.1 1, 2 461
GenBank mRNA AK009942.1 - 1, 2 302
BC005568.1 - 1, 2 461 3967350

View Full Product Details