Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323956.1 | NP_001310885.1 | 10 | 1907 | |
NM_001323958.1 | NP_001310887.1 | 9 | 1762 | ||
NM_006462.5 | NP_006453.1 | 10 | 1923 | ||
NM_031229.3 | NP_112506.2 | 11 | 2068 | ||
NR_136659.1 | 11 | 2043 | |||
XM_005260645.1 | XP_005260702.1 | 11 | 1864 | ||
XM_011529137.1 | XP_011527439.1 | 12 | 1987 | ||
XM_011529138.1 | XP_011527440.1 | 12 | 1942 | ||
XM_011529139.2 | XP_011527441.1 | 12 | 2010 | ||
XM_017027594.1 | XP_016883083.1 | 10 | 1952 | ||
XM_017027595.1 | XP_016883084.1 | 10 | 1297 | ||
GenBank mRNA | AB107766.1 | - | 10 | 1254 | |
BC000983.2 | - | 9 | 1457 | ||
BC015219.2 | - | 11 | 1781 | 3877587 | |
U67322.1 | - | 10 | 1809 |