Gene Transcripts

Gene Symbol
STAG2
Entrez Gene ID
10735
Gene Name
stromal antigen 2
Gene Aliases
SA-2, SA2, SCC3B, bA517O1.1
Chromosome Location
Chr. X: 123960560 - 124102656 on Build GRCh38
UniGene ID
Hs.496710
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042749.2 NP_001036214.1 30 3706
NM_001042750.1 NP_001036215.1 30 3550
NM_001042751.1 NP_001036216.1 30 3550
NM_001282418.1 NP_001269347.1 30 3529
NM_006603.4 NP_006594.3 29 3485
XM_005262357.2 XP_005262414.1 29 3575
XM_005262358.2 XP_005262415.1 30 3512
XM_005262359.3 XP_005262416.1 29 3465
XM_005262360.2 XP_005262417.1 29 3394
XM_005262361.2 XP_005262418.1 29 3522
XM_006724727.1 XP_006724790.1 31 3654
XM_006724728.3 XP_006724791.1 30 3588
XM_011531253.2 XP_011529555.1 29 3447
XM_017029232.1 XP_016884721.1 30 3512
XM_017029233.1 XP_016884722.1 29 3394
XM_017029234.1 XP_016884723.1 29 3447
XM_017029235.1 XP_016884724.1 30 3593
GenBank mRNA AK298633.1 - 19 2204
AK313559.1 - 29 3377
AL831939.2 - 29 3453
BC001765.1 - 29 3434 3352347
BX640970.1 - 30 3561
BX641002.1 - 30 3454
BX641003.1 - 30 3519
CR627425.1 - 30 3528
CR749433.1 - 30 3519

View Full Product Details