Gene Transcripts

Gene Symbol
Glp2r
Entrez Gene ID
93896
Gene Name
glucagon-like peptide 2 receptor
Gene Aliases
9530092J08Rik, GLP-2
Chromosome Location
Chr. 11: 67663299 - 67771153 on Build GRCm38
UniGene ID
Mm.118883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175681.3 NP_783612.2 11 1390
GenBank mRNA AK035702.1 - 785
AY605231.1 - 11 1109
BC044746.2 - 11 1390 5363416

View Full Product Details