Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006656.5 | NP_006647.3 | 3 | 1283 | |
GenBank mRNA | AB008185.2 | - | 4 | 1198 | |
AK022450.1 | - | 3 | 1277 | ||
AK290442.1 | - | 3 | 1245 | ||
AK303540.1 | - | 2 | 1031 | ||
BC136397.1 | - | 4 | 1617 | 9020384 | |
BC144059.1 | - | 4 | 1617 | 9052579 | |
Y18563.1 | - | 3 | 2054 |