Gene Transcripts

Gene Symbol
C1R
Entrez Gene ID
715
Gene Name
complement C1r subcomponent
Gene Aliases
Chromosome Location
Chr. 12: 7080209 - 7092447 on Build GRCh38
UniGene ID
Hs.524224
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001733.4 NP_001724.3 3 444
XM_017019915.1 XP_016875404.1 3 518
GenBank mRNA AK024951.1 - 2 1144
AK222481.1 - 3 438
AK222491.1 - 3 438
AK291313.1 - 3 466
AK298445.1 - 3 459
AK303750.1 - 3 337
BC035220.1 - 3 444 4777940
M14058.1 - 3 448
X04701.1 - 3 436

View Full Product Details