Gene Transcripts

Gene Symbol
HNRNPA0
Entrez Gene ID
10949
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Gene Aliases
HNRPA0
Chromosome Location
Chr. 5: 137751384 - 137754350 on Build GRCh38
UniGene ID
Hs.645902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006805.3 NP_006796.1 1 1805
GenBank mRNA AA081021.1 - 1 176
BC018949.2 - 1 1805 4131137

View Full Product Details