Gene Transcripts

Gene Symbol
TLK2
Entrez Gene ID
11011
Gene Name
tousled like kinase 2
Gene Aliases
HsHPK, PKU-ALPHA
Chromosome Location
Chr. 17: 62477481 - 62615481 on Build GRCh38
UniGene ID
Hs.445078
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284333.1 NP_001271262.1 18 1807
NM_001284363.1 NP_001271292.1 15 1402
NM_006852.3 NP_006843.2 17 1744
XM_011524215.2 XP_011522517.1 16 1451
XM_011524216.2 XP_011522518.1 18 1571
XM_011524217.2 XP_011522519.1 18 1758
XM_011524218.2 XP_011522520.1 15 1385
XM_011524220.2 XP_011522522.1 15 1355
XM_011524221.2 XP_011522523.1 17 1469
XM_011524222.2 XP_011522524.1 18 1950
XM_011524223.2 XP_011522525.1 16 1612
XM_011524224.2 XP_011522526.1 16 1670
XM_011524225.2 XP_011522527.1 16 1618
XM_011524226.2 XP_011522528.1 18 1664
XM_011524227.2 XP_011522529.1 17 1718
XM_011524228.2 XP_011522530.1 18 1818
XM_011524229.2 XP_011522531.1 19 1745
XM_017024044.1 XP_016879533.1 17 1578
XM_017024045.1 XP_016879534.1 16 1686
XM_017024046.1 XP_016879535.1 17 1508
XM_017024047.1 XP_016879536.1 17 1505
XM_017024048.1 XP_016879537.1 17 1966
XM_017024049.1 XP_016879538.1 17 1919
XM_017024050.1 XP_016879539.1 15 1451
XM_017024051.1 XP_016879540.1 16 1900
XM_017024052.1 XP_016879541.1 16 1851
XM_017024053.1 XP_016879542.1 16 1523
XM_017024054.1 XP_016879543.1 15 1549
XM_017024055.1 XP_016879544.1 14 1453
XM_017024056.1 XP_016879545.1 15 1604
XM_017024057.1 XP_016879546.1 15 1418
XM_017024058.1 XP_016879547.1 17 1696
XM_017024059.1 XP_016879548.1 17 1686
XM_017024060.1 XP_016879549.1 16 1603
GenBank mRNA AB004884.1 - 15 1380
AF162667.1 - 17 1616
AK300586.1 - 15 1402
AX775745.1 - 17 1616
BC044925.1 - 18 1714

View Full Product Details