Gene Transcripts

Gene Symbol
RAB31
Entrez Gene ID
11031
Gene Name
RAB31, member RAS oncogene family
Gene Aliases
Rab22B
Chromosome Location
Chr. 18: 9707971 - 9862556 on Build GRCh38
UniGene ID
Hs.744887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006868.3 NP_006859.2 6 588
XM_017025528.1 XP_016881017.1 7 970
XM_017025529.1 XP_016881018.1 7 958
GenBank mRNA AF183421.1 - 6 541
AF234995.1 - 6 587
AF498957.1 - 6 410
AK223505.1 - 6 452
AK314809.1 - 6 546
BC001148.1 - 6 510 3534853
BT020027.1 - 6 410
BT020028.1 - 6 410
BU944638.1 - 6 654
CR407659.1 - 6 410
U57091.1 - 6 455
U59877.1 - 6 470

View Full Product Details