Gene Transcripts
- Gene Symbol
- Avp
- Entrez Gene ID
- 24221
- Gene Name
- arginine vasopressin
- Gene Aliases
- DI, VP, Vas
- Chromosome Location
- Chr. 3: 123117482 - 123119460 on Build Rnor_6.0
- UniGene ID
- Rn.9976
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016992.2 |
NP_058688.2
|
1 |
106 |
|
GenBank mRNA |
M25646.1 |
- |
1 |
106 |
|
View Full Product Details