Gene Transcripts

Gene Symbol
Zscan4b
Entrez Gene ID
665780
Gene Name
zinc finger and SCAN domain containing 4B
Gene Aliases
EG665780
Chromosome Location
Chr. 7: 10900740 - 10905050 on Build GRCm38
UniGene ID
Mm.482067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185173.1 NP_001172102.1 4 1580

View Full Product Details