Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001106296.2 | NP_001099766.2 | 3 | 470 | |
XM_006230237.2 | XP_006230299.1 | 4 | 546 | ||
XM_006230239.2 | XP_006230301.1 | 3 | 291 | ||
XM_006230240.2 | XP_006230302.1 | 3 | 1066 | ||
GenBank mRNA | BC161920.1 | - | 3 | 470 |