Gene Transcripts

Gene Symbol
NISCH
Entrez Gene ID
11188
Gene Name
nischarin
Gene Aliases
I-1, IR1, IRAS, hIRAS
Chromosome Location
Chr. 3: 52455508 - 52493072 on Build GRCh38
UniGene ID
Hs.435290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007184.3 NP_009115.2 13, 14 1671
XM_005264839.2 XP_005264896.1 12, 13 1520
XM_017005643.1 XP_016861132.1 4, 5 512
GenBank mRNA AB023192.1 - 14, 15 1609
AF058290.1 - 1, 2 134
AF082516.1 - 13, 14 1563
AK027428.1 - 13, 14 1549
AK291398.1 - 13, 14 1575
AK302441.1 - 4, 5 559
AK315001.1 - 13, 14 1575
AL117432.1 - 1 130
BC038102.2 - 13, 14 1575 5268247
BC054494.1 - 13, 14 1569 6195532
BC056900.1 - 13, 14 1541 6143316

View Full Product Details