Gene Transcripts

Gene Symbol
Sh3pxd2b
Entrez Gene ID
303021
Gene Name
SH3 and PX domains 2B
Gene Aliases
RGD1309926
Chromosome Location
Chr. 10: 17209152 - 17296449 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246109.2 XP_006246171.1 6 473
XM_008767621.1 XP_008765843.1 5 1044
XM_008767622.1 XP_008765844.1 5 1044

View Full Product Details