Gene Transcripts

Gene Symbol
Gpc6
Entrez Gene ID
691984
Gene Name
glypican 6
Gene Aliases
Chromosome Location
Chr. 15: 102164183 - 103174721 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001077762.5 XP_001077762.3 9 3000
XM_003751533.3 XP_003751581.1 10 3031

View Full Product Details