Gene Transcripts

Gene Symbol
Spef1
Entrez Gene ID
311429
Gene Name
sperm flagellar 1
Gene Aliases
RGD1311267
Chromosome Location
Chr. 3: 123714564 - 123720786 on Build Rnor_6.0
UniGene ID
Rn.19612
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039024.2 NP_001034113.2 3 452
XM_006235033.2 XP_006235095.1 3 2806
GenBank mRNA BC099127.1 - 3 452

View Full Product Details