Gene Transcripts

Gene Symbol
Rasgef1a
Entrez Gene ID
312664
Gene Name
RasGEF domain family, member 1A
Gene Aliases
Chromosome Location
Chr. 4: 150064181 - 150144203 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006237211.2 XP_006237273.1 2 224
XM_008763269.1 XP_008761491.1 2 360
XM_232315.7 XP_232315.6 2 386

View Full Product Details