Gene Transcripts

Gene Symbol
Rtp4
Entrez Gene ID
360733
Gene Name
receptor (chemosensory) transporter protein 4
Gene Aliases
RGD1306974
Chromosome Location
Chr. 11: 80638945 - 80650802 on Build Rnor_6.0
UniGene ID
Rn.4244
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108321.1 NP_001101791.1 3 349
XM_006248521.2 XP_006248583.1 1 736

View Full Product Details