Gene Transcripts

Gene Symbol
Cnpy4
Entrez Gene ID
363886
Gene Name
canopy FGF signaling regulator 4
Gene Aliases
Cnpy, RGD1307636
Chromosome Location
Chr. 12: 19329027 - 19335126 on Build Rnor_6.0
UniGene ID
Rn.6578
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108852.2 NP_001102322.1 2 174
GenBank mRNA BC162041.1 - 2 174
FQ226241.1 - 2 193
FQ229096.1 - 2 219

View Full Product Details