Gene Transcripts

Gene Symbol
Mcpt1l2
Entrez Gene ID
408238
Gene Name
mast cell protease 1-like 2
Gene Aliases
Chromosome Location
Chr. 15: 34633988 - 34636186 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252034.2 XP_006252096.1 2 109

Gene Symbol
LOC498518
Entrez Gene ID
498518
Gene Name
hypothetical LOC498518
Gene Aliases
Chromosome Location
Chr. 15: 34680100 - 34691247 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770726.1 XP_008768948.1 2 279

View Full Product Details