Gene Transcripts

Gene Symbol
Cbln1
Entrez Gene ID
498922
Gene Name
cerebellin 1 precursor
Gene Aliases
Chromosome Location
Chr. 19: 20607507 - 20611316 on Build Rnor_6.0
UniGene ID
Rn.24402
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109127.1 NP_001102597.1 3 906
GenBank mRNA BC087589.1 - 1 508

View Full Product Details