Gene Transcripts

Gene Symbol
Vom2r80
Entrez Gene ID
502285
Gene Name
vomeronasal 2 receptor, 80
Gene Aliases
RGD1559687
Chromosome Location
Chr. 1: 62500357 - 62508149 on Build Rnor_6.0
UniGene ID
Rn.218962
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111321.1 NP_001104791.1 782

View Full Product Details