Gene Transcripts
- Gene Symbol
- Sval1
- Entrez Gene ID
- 680174
- Gene Name
- seminal vesicle antigen-like 1
- Gene Aliases
- SLP-C
- Chromosome Location
- Chr. 4: 71376535 - 71383953 on Build Rnor_6.0
- UniGene ID
- Rn.220735
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133292.2 |
NP_579826.1
|
2 |
166 |
|
View Full Product Details